siRNA Details
| Gene Symbol: D830032E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102306.1 | 4 | 575 | |||
| NR_102307.1 | 5 | 689 | ||||
| GenBank mRNA | AK085905.1 | - | 4 | 576 | ||
| AK085943.1 | - | 5 | 664 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB662147.1 | - | 4 | 576 | Chr. 1 - 107917259 - 107949469 [+] on Build GRCm38 | |

D830032E09Rik
Genomic Map