siRNA Details
| Gene Symbol: Gm17751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038012.1 | 5 | 770 | |||
| NR_038013.1 | 5 | 586 | ||||
| GenBank mRNA | AK054265.1 | - | 5 | 586 | ||
| AK162144.1 | - | 5 | 770 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CO807055.1 | - | 5 | 560 | Chr. 1 - 76489159 - 76500833 [+] on Build GRCm38 | |

Gm17751
Genomic Map