siRNA Details
| Gene Symbol: 1700120C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045627.2 | 1 | 91 | |||
| GenBank mRNA | AK007210.1 | - | 1 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700120C14Rik| Gene Symbol: 1700120C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045627.2 | 1 | 91 | |||
| GenBank mRNA | AK007210.1 | - | 1 | 92 | ||