siRNA Details
| Gene Symbol: A230077H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040329.1 | 4 | 496 | |||
| GenBank mRNA | AK038939.1 | - | 1019 | |||
| AK081136.1 | - | 5 | 497 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230077H06Rik| Gene Symbol: A230077H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040329.1 | 4 | 496 | |||
| GenBank mRNA | AK038939.1 | - | 1019 | |||
| AK081136.1 | - | 5 | 497 | |||