siRNA Details
| Gene Symbol: 9430060I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015525.1 | 1, 2 | 113 | |||
| GenBank mRNA | BC090260.1 | - | 1, 2 | 113 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ922611.1 | - | 112 | Chr. - - [-] on Build | ||

9430060I03Rik
Genomic Map