siRNA Details
| Gene Symbol: 1700083H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006983.1 | - | 2 | 407 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707040.1 | - | 2 | 407 | Chr. 12 - 71383154 - 71384208 [-] on Build GRCm38 | |

1700083H02Rik
Genomic Map