siRNA Details
| Gene Symbol: 1700120G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046050.1 | 2 | 109 | |||
| GenBank mRNA | AK007216.1 | - | 2 | 110 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707307.1 | - | 2 | 113 | Chr. 7 - 135191765 - 135196002 [-] on Build GRCm38 | |

1700120G07Rik
Genomic Map