siRNA Details
| Gene Symbol: 2010106C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045435.1 | 3 | 613 | |||
| GenBank mRNA | AK008318.1 | - | 3 | 615 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010106C02Rik| Gene Symbol: 2010106C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045435.1 | 3 | 613 | |||
| GenBank mRNA | AK008318.1 | - | 3 | 615 | ||