siRNA Details
Gene Symbol: 1700041M19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040573.1 | 2 | 367 | |||
GenBank mRNA | AK006678.1 | - | 2 | 368 | ||
AK077115.1 | - | 2 | 362 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700041M19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040573.1 | 2 | 367 | |||
GenBank mRNA | AK006678.1 | - | 2 | 368 | ||
AK077115.1 | - | 2 | 362 |