siRNA Details
| Gene Symbol: 1700041M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040573.1 | 2 | 209 | |||
| GenBank mRNA | AK006678.1 | - | 2 | 210 | ||
| AK077115.1 | - | 2 | 204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700041M19Rik| Gene Symbol: 1700041M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040573.1 | 2 | 209 | |||
| GenBank mRNA | AK006678.1 | - | 2 | 210 | ||
| AK077115.1 | - | 2 | 204 | |||