siRNA Details
| Gene Symbol: 2210022D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008767.1 | - | 2 | 284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210022D18Rik| Gene Symbol: 2210022D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008767.1 | - | 2 | 284 | ||