siRNA Details
Gene Symbol: 1700030N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045304.1 | 3 | 323 | |||
GenBank mRNA | AK007025.1 | - | 3 | 324 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700030N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045304.1 | 3 | 323 | |||
GenBank mRNA | AK007025.1 | - | 3 | 324 |