siRNA Details
| Gene Symbol: Gm10640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046062.1 | 1 | 194 | |||
| GenBank mRNA | AK133438.1 | - | 1 | 194 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ047870.1 | - | 1 | 194 | Chr. 7 - 31162358 - 31163934 [+] on Build GRCm38 | |

Gm10640
Genomic Map