siRNA Details
| Gene Symbol: 1700100L14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045934.1 | 3 | 426 | |||
| GenBank mRNA | AK007102.1 | - | 3 | 428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700100L14Rik| Gene Symbol: 1700100L14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045934.1 | 3 | 426 | |||
| GenBank mRNA | AK007102.1 | - | 3 | 428 | ||