siRNA Details
| Gene Symbol: Gm10538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045892.1 | 1 | 561 | |||
| GenBank mRNA | AK133041.1 | - | 1 | 562 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10538| Gene Symbol: Gm10538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045892.1 | 1 | 561 | |||
| GenBank mRNA | AK133041.1 | - | 1 | 562 | ||