siRNA Details
| Gene Symbol: 2010310C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045169.1 | 4 | 550 | |||
| GenBank mRNA | AK008560.1 | - | 4 | 552 | ||
| AK028457.1 | - | 1275 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010310C07Rik| Gene Symbol: 2010310C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045169.1 | 4 | 550 | |||
| GenBank mRNA | AK008560.1 | - | 4 | 552 | ||
| AK028457.1 | - | 1275 | ||||