siRNA Details
| Gene Symbol: BC055402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037990.1 | 4, 5 | 836 | |||
| GenBank mRNA | AK149264.1 | - | 4, 5 | 727 | ||
| BC055402.1 | - | 4, 5 | 833 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC055402| Gene Symbol: BC055402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037990.1 | 4, 5 | 836 | |||
| GenBank mRNA | AK149264.1 | - | 4, 5 | 727 | ||
| BC055402.1 | - | 4, 5 | 833 | |||