siRNA Details
Gene Symbol: BC055402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037990.1 | 2 | 302 | |||
GenBank mRNA | AK149264.1 | - | 2 | 193 | ||
BC055402.1 | - | 2 | 299 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC055402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037990.1 | 2 | 302 | |||
GenBank mRNA | AK149264.1 | - | 2 | 193 | ||
BC055402.1 | - | 2 | 299 |