siRNA Details
| Gene Symbol: 1700109G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033788.1 | 2 | 584 | |||
| GenBank mRNA | AK007149.1 | - | 2 | 584 | ||
| AK018935.1 | - | 2 | 547 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700109G14Rik| Gene Symbol: 1700109G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033788.1 | 2 | 584 | |||
| GenBank mRNA | AK007149.1 | - | 2 | 584 | ||
| AK018935.1 | - | 2 | 547 | |||