siRNA Details
Gene Symbol: 1700109G14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033788.1 | 1 | 134 | |||
GenBank mRNA | AK007149.1 | - | 1 | 134 | ||
AK007289.1 | - | 1 | 109 | |||
AK018935.1 | - | 1 | 97 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700109G14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033788.1 | 1 | 134 | |||
GenBank mRNA | AK007149.1 | - | 1 | 134 | ||
AK007289.1 | - | 1 | 109 | |||
AK018935.1 | - | 1 | 97 |