siRNA Details
| Gene Symbol: 1700112H15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040472.1 | 6 | 634 | |||
| GenBank mRNA | AK007177.1 | - | 6 | 642 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700112H15Rik| Gene Symbol: 1700112H15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040472.1 | 6 | 634 | |||
| GenBank mRNA | AK007177.1 | - | 6 | 642 | ||