siRNA Details
| Gene Symbol: 1600010M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037959.1 | 2, 3 | 227 | |||
| GenBank mRNA | AK005418.1 | - | 848 | |||
| AK162666.1 | - | 2, 3 | 228 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1600010M07Rik| Gene Symbol: 1600010M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037959.1 | 2, 3 | 227 | |||
| GenBank mRNA | AK005418.1 | - | 848 | |||
| AK162666.1 | - | 2, 3 | 228 | |||