siRNA Details
| Gene Symbol: Gm29371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077157.1 | - | 2 | 620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm29371| Gene Symbol: Gm29371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077157.1 | - | 2 | 620 | ||