siRNA Details
| Gene Symbol: 4930579H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016323.1 | - | 5 | 449 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715962.1 | - | 5 | 449 | Chr. 10 - 27389866 - 27396480 [+] on Build GRCm38 | |

4930579H20Rik
Genomic Map