siRNA Details
| Gene Symbol: 1700094J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040580.1 | 3 | 566 | |||
| GenBank mRNA | AK007067.1 | - | 3 | 565 | ||
| AK019782.1 | - | 1 | 108 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715828.1 | - | 1 | 108 | Chr. 10 - 76398593 - 76398968 [+] on Build GRCm38 | |

1700094J05Rik
Genomic Map