siRNA Details
| Gene Symbol: 4930521E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040602.1 | 3 | 839 | |||
| GenBank mRNA | AK015860.1 | - | 3 | 840 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930521E06Rik| Gene Symbol: 4930521E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040602.1 | 3 | 839 | |||
| GenBank mRNA | AK015860.1 | - | 3 | 840 | ||