siRNA Details
| Gene Symbol: Gm28979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110509.1 | 3 | 901 | |||
| GenBank mRNA | AK043886.1 | - | 3 | 902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm28979| Gene Symbol: Gm28979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110509.1 | 3 | 901 | |||
| GenBank mRNA | AK043886.1 | - | 3 | 902 | ||