siRNA Details
| Gene Symbol: Gm20125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038020.1 | 9 | 1565 | |||
| GenBank mRNA | AK029503.1 | - | 9 | 1567 | ||
| AK136184.1 | - | 3 | 535 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720160.1 | - | 3 | 531 | Chr. 10 - 16952789 - 16956010 [-] on Build GRCm38 | |

Gm20125
Genomic Map