siRNA Details
| Gene Symbol: 4933400F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015540.1 | 1, 2 | 214 | |||
| GenBank mRNA | AK007030.1 | - | 1, 2 | 211 | ||
| AK016229.1 | - | 233 | ||||
| AK016592.1 | - | 1, 2 | 216 | |||
| AK029704.1 | - | 1, 2 | 218 | |||

4933400F21Rik
Genomic Map