siRNA Details
| Gene Symbol: Gm5092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030245.1 | - | 3 | 173 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB614465.1 | - | 3 | 173 | Chr. 17 - 21091208 - 21098539 [+] on Build GRCm38 | |

Gm5092
Genomic Map