siRNA Details
| Gene Symbol: 1810041H14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019031.1 | - | 2 | 334 | ||
| BY707761.1 | - | 2 | 334 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA462605.1 | - | 371 | Chr. - - [-] on Build | ||

1810041H14Rik
Genomic Map