siRNA Details
| Gene Symbol: C2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104191.1 | 1 | 296 | |||
| GenBank mRNA | BX100594.1 | - | 1 | 295 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000630806.1 | - | 296 | Chr. - - [-] on Build | ||

C2-AS1
Genomic Map