siRNA Details
Gene Symbol: JAKMIP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038902.1 | 4 | 652 | |||
GenBank mRNA | AK027578.1 | - | 4 | 652 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000626945.2 | - | 652 | Chr. - - [-] on Build |
