siRNA Details
| Gene Symbol: JAKMIP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038902.1 | 4 | 1355 | |||
| GenBank mRNA | AK027578.1 | - | 4 | 1355 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000626945.2 | - | 1355 | Chr. - - [-] on Build | ||

JAKMIP2-AS1
Genomic Map