siRNA Details
| Gene Symbol: LINC01127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055877.1 | - | 3 | 2583 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000627215.1 | - | 2583 | Chr. - - [-] on Build | ||

LINC01127
Genomic Map