siRNA Details
| Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024496.1 | 2 | 3002 | |||
| NR_135010.1 | 3 | 2421 | ||||
| GenBank mRNA | BC068605.1 | - | 3 | 2423 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000619442.1 | - | 2420 | Chr. - - [-] on Build | ||

PTGER4P2-CDK2AP2P2
Genomic Map