siRNA Details
Gene Symbol: PCAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110138.1 | 3 | 2228 | |||
GenBank mRNA | KF154780.1 | - | 3 | 2228 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000626667.1 | - | 2228 | Chr. - - [-] on Build |
