siRNA Details
| Gene Symbol: PCAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110138.1 | 2 | 624 | |||
| GenBank mRNA | KF154780.1 | - | 2 | 624 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000626667.1 | - | 624 | Chr. - - [-] on Build | ||

PCAT5
Genomic Map