siRNA Details
Gene Symbol: LOC101929297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125862.1 | 2, 3 | 1205 | |||
GenBank mRNA | BC040873.1 | - | 2, 3 | 1205 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000444219.2 | - | 1205 | Chr. 6 - 166236969 - 166243709 [+] on Build GRCh38 |
