siRNA Details
| Gene Symbol: LINC00506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047469.1 | 2 | 4035 | |||
| GenBank mRNA | BC030609.1 | - | 2 | 4038 | ||
| BC050344.1 | - | 2 | 4033 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000629295.1 | - | 4035 | Chr. - - [-] on Build | ||

LINC00506
Genomic Map