siRNA Details
| Gene Symbol: LINC00506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047469.1 | 1 | 21 | |||
| NR_104153.1 | 1 | 21 | ||||
| GenBank mRNA | BC030609.1 | - | 1 | 24 | ||
| BC050344.1 | - | 1 | 19 | |||
| DA227977.1 | - | 1 | 173 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00506| Gene Symbol: LINC00506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047469.1 | 1 | 21 | |||
| NR_104153.1 | 1 | 21 | ||||
| GenBank mRNA | BC030609.1 | - | 1 | 24 | ||
| BC050344.1 | - | 1 | 19 | |||
| DA227977.1 | - | 1 | 173 | |||