siRNA Details
| Gene Symbol: LOC101927079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110480.1 | 8, 9 | 1105 | |||
| NR_110481.1 | 6, 7 | 837 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000638243.1 | - | 837 | Chr. - - [-] on Build | ||

LOC101927079
Genomic Map