siRNA Details
Gene Symbol: MIR670HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131246.1 | 1 | 93 | |||
GenBank mRNA | BC031305.2 | - | 1 | 93 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000533531.1 | - | 1 | 93 | Chr. 11 - 43569306 - 43570395 [+] on Build GRCh38 |
