siRNA Details
| Gene Symbol: LINC01241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121604.1 | 6 | 984 | |||
| GenBank mRNA | BC043546.1 | - | 6 | 984 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000423326.2 | - | 984 | Chr. 9 - 25780056 - 25784562 [+] on Build GRCh38 | ||

LINC01241
Genomic Map