siRNA Details
Gene Symbol: LOC101927179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125880.1 | 2 | 255 | |||
NR_125881.1 | 2 | 226 | ||||
GenBank mRNA | AL599247.1 | - | 2 | 227 | ||
BX280044.1 | - | 2 | 217 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125880.1 | 2 | 255 | |||
NR_125881.1 | 2 | 226 | ||||
GenBank mRNA | AL599247.1 | - | 2 | 227 | ||
BX280044.1 | - | 2 | 217 |