siRNA Details
| Gene Symbol: LOC101927179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125880.1 | 3 | 366 | |||
| NR_125881.1 | 3 | 337 | ||||
| GenBank mRNA | AL599247.1 | - | 3 | 338 | ||
| BX280044.1 | - | 3 | 328 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927179| Gene Symbol: LOC101927179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125880.1 | 3 | 366 | |||
| NR_125881.1 | 3 | 337 | ||||
| GenBank mRNA | AL599247.1 | - | 3 | 338 | ||
| BX280044.1 | - | 3 | 328 | |||