siRNA Details
| Gene Symbol: LOC100996385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103794.1 | 3 | 1045 | |||
| GenBank mRNA | BC042064.1 | - | 3 | 1012 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000512675.1 | - | 3 | 1011 | Chr. 5 - 176049678 - 176062021 [-] on Build GRCh38 | |

LOC100996385
Genomic Map