siRNA Details
| Gene Symbol: LOC101929733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110575.1 | 2 | 371 | |||
| GenBank mRNA | BG201257.1 | - | 2 | 368 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000607419.1 | - | 2 | 369 | Chr. 2 - 10878269 - 10885118 [+] on Build GRCh38 | |

LOC101929733
Genomic Map