siRNA Details
| Gene Symbol: FAM87B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103536.1 | 2 | 1185 | |||
| GenBank mRNA | AK097327.1 | - | 2 | 1185 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000326734.2 | - | 2 | 1185 | Chr. 1 - 817371 - 819834 [+] on Build GRCh38 | |

FAM87B
Genomic Map