siRNA Details
| Gene Symbol: LOC105377924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134603.1 | 2 | 241 | |||
| GenBank mRNA | BX105858.1 | - | 2 | 241 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000632749.1 | - | 241 | Chr. - - [-] on Build | ||

LOC105377924
Genomic Map